Learn More
DWK Life Sciences Wheaton™ AntiBind™ 384 Deep Well Microplates, Conical, 120μL
Used in chromatography applications, long term sample storage, and bioanalytical assays. Wheaton AntiBIND 384 Deep Well Microplates, Conical, 120μL are manufactured using a new technology that modifies the polypropylene surface at the molecular level to produce a homogenous hydrophilic surface that eliminates nonspecific protein binding and ensures ultimate recovery of valuable proteins.
Supplier:  DWK Life Sciences W266042
 View more versions of this product
Catalog No. 02-911-705
Description
Description
  • Application of Advanced Plasma Treatment technology (APT) allows for comprehensive complete hydrophilic treatment that is not compromised in presence of solvents
  • Meets the SBS and ANSI dimensions used for automation
  • Conical shaped bottoms for maximum sample recovery and efficient mixing
  • Manufactured from polypropylene resin
Specifications
Specifications
16 x 24
High through-put drug discovery, proteomics research, bioanalytical assays, long term sample storage, sample prep, chromatography (LC-MS)
120
AntiBIND
Standard
Natural
Polypropylene
120/Cs.
384
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.