Learn More
National Safety Apparel™ ArcGuard™ Compliance 40 Cal Long Coat Kit (10)
SDP
With a unique blend of 88% cotton and 12% high-tenacity nylon UltraSoft garments provide a smooth feel for increased comfort and the inherently FR material delivers exceptional performance in reliability
$1909.00 - $2079.00
Specifications
Ankle Style Open
Arc Flash Rating 40 cal/cm²
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269
Closure Type FR Hook and Loop
Color Navy
View More Specs
Products 8
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-001-1464
SDP
View Documents
National Safety Apparel™
KIT4LC40SM10
Small
Each for $1,909.00
missing translation for 'QuantityLeft'
Add to Cart
17-001-1469
SDP
View Documents
National Safety Apparel™
KIT4LC40MD10
Medium
Each for $1,909.00
missing translation for 'QuantityLeft'
Add to Cart
17-001-1474
SDP
View Documents
National Safety Apparel™
KIT4LC40LG10
Large
Each for $1,948.00
missing translation for 'QuantityLeft'
Add to Cart
17-001-1479
SDP
View Documents
National Safety Apparel™
KIT4LC40XL10
X-Large
Each for $1,909.00
missing translation for 'QuantityLeft'
Add to Cart
17-001-1484
SDP
View Documents
National Safety Apparel™
KIT4LC402X10
2X-Large
Each for $1,962.00
missing translation for 'QuantityLeft'
Add to Cart
17-001-1489
SDP
View Documents
National Safety Apparel™
KIT4LC403X10
3X-Large
Each for $1,962.00
missing translation for 'QuantityLeft'
Add to Cart
17-001-1494
SDP
View Documents
National Safety Apparel™
KIT4LC404X10
4X-Large
Each for $2,015.00
missing translation for 'QuantityLeft'
Add to Cart
17-001-1499
SDP
View Documents
National Safety Apparel™
KIT4LC405X10
5X-Large
Each for $2,079.00
missing translation for 'QuantityLeft'
Add to Cart
Description
Description
  • Arc Rating: 40 cal/cm²
  • CAT 4
  • Fabric/Material: 9 oz. Westex™ UltraSoft™ Sateen, Double Layer
  • FR hook and loop front closure
  • Stand up collar for extra coverage
  • Rib knit cuffs for comfortable, secure fit
  • Garments embroidered with arc rating
  • Quick disconnect and adjustable shoulder straps on bib overalls
  • Expandable leg opening to fit over boots
  • Made in the USA
Specifications
Specifications
Open
CAT 4, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269
Navy
Stitched
40 cal/cm²
FR Hook and Loop
Cotton, Nylon, Polyester
Adjustable
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.