Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Phoenix™ Becoming Banneker Kit Components
SCI_ED

Replacement parts for Educational Kit

$.38 - $19.62

Specifications

Grade K to 5
For Use With (Application) Education Kit
View More Specs

Products 10
Catalog Number Product Type Price Quantity & Availability  
Catalog Number Product Type Price Quantity & Availability  
S396205BB
SCI_ED
View Documents
Plastic bags
Each for $19.62
missing translation for 'QuantityLeft'
Add to Cart
 
S396215BB
SCI_ED
View Documents
Foam cube
Each for $2.80
missing translation for 'QuantityLeft'
Add to Cart
 
S396225BB
SCI_ED
View Documents
Keychain flashlight
Each for $10.05
missing translation for 'QuantityLeft'
Add to Cart
 
S396235BB
SCI_ED
View Documents
Flashlight
Each for $6.72
missing translation for 'QuantityLeft'
Add to Cart
 
S396245BB
SCI_ED
View Documents
Sidewalk chalk
Each for $3.71
missing translation for 'QuantityLeft'
Add to Cart
 
S396255BB
SCI_ED
View Documents
Clear Plastic Protractor
Each for $0.70
missing translation for 'QuantityLeft'
Add to Cart
 
S396265BB
SCI_ED
View Documents
Brass Fasteners
Each for $18.01
missing translation for 'QuantityLeft'
Add to Cart
 
S396275BB
SCI_ED
View Documents
Glow in the Dark Star Finder
Each for $15.86
missing translation for 'QuantityLeft'
Add to Cart
 
S396285BB
SCI_ED
View Documents
Adding Machine Paper
Each for $1.88
missing translation for 'QuantityLeft'
Add to Cart
 
S396295BB
SCI_ED
View Documents
Cardstock
Each for $0.38
missing translation for 'QuantityLeft'
Add to Cart
 
Description

Description

Variety of components specially formatted as replacements for AquaPhoenix custom designed hands-on educational kits.
Specifications

Specifications

K to 5
Education Kit
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.