Learn More
STERIS™ Cage-Klenz 220 Acid-Based Detergent
Cage-Klenz 220 Acid-Based Detergent is a specially formulated glycolic acid detergent used for the safe and effective removal of hard water scale, urine scale and organic residues found in research animal caging. pH (1%): 2.8, typical.
Supplier: STERIS™ 1K22D3
Description
Cage-Klenz 220 Detergent is specially formulated for the safe and effective removal of hard water scale, urine scale and organic residues found in research animal caging.
HOW CAGE-KLENZ 220 DETERGENT WORKS
Cage-Klenz 220 Detergent is a liquid phosphate-free, hydroxyacetic acid-based detergent.
WHY CAGE-KLENZ 220 DETERGENT?
- Efficacy - Effectively cleans hard water and urine scale, and organic soils in both soft and hard water
- Flexibility - Excellent materials compatibility with a wide variety of caging materials including polycarbonate
- Cost Savings - Reduces utility consumption and cleaning time due with low foaming, free-rinsing formulation
- Environmentally Friendly - phosphate-free formulation with biodegradable surfactants
- Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility
pH (1%): 2.8, typical
Specifications
Acid Cleaner | |
Clear Liquid | |
Unscented | |
Keg |
No | |
2.8 (Diluted) | |
Non-sterile | |
30 gal. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.