Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonic Anhydrase VI Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5145389

Description
The protein encoded by this gene is one of several isozymes of carbonic anhydrase. This protein is found only in salivary glands and saliva and protein may play a role in the reversible hydratation of carbon dioxide though its function in saliva is unknown.Specifications
Carbonic Anhydrase VI | |
Polyclonal | |
Unconjugated | |
Car6 | |
Car6 | |
Recombinant Rat CA6 protein, Thr147-Ala275 (Accession #F1LQ08) | |
200 μL | |
Primary | |
Rat | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% Proclin 300, pH 7.4 | |
CA6, CA-6, Car6, Carbonate dehydratase VI, carbonic anhydrase 6, Carbonic anhydrase VI, carbonic anhydrase VI nirs variant 2, Carbonic anhydrase6, CA-VI, DOC1, GUSTIN, salivary carbonic anhydrase, Secreted carbonic anhydrase | |
Rabbit | |
Protein A | |
RUO | |
298657 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction