Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonic Anhydrase VI Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
$178.80 - $408.00
Specifications
Antigen | Carbonic Anhydrase VI |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEPA5145390
![]() |
Thermo Scientific
PA5145390 |
20 μL |
Each for $178.80
|
|
|||||
FEPA5145389
![]() |
Thermo Scientific
PA5145389 |
200 μL |
Each for $408.00
|
|
|||||
Description
The protein encoded by this gene is one of several isozymes of carbonic anhydrase. This protein is found only in salivary glands and saliva and protein may play a role in the reversible hydratation of carbon dioxide though its function in saliva is unknown.Specifications
Carbonic Anhydrase VI | |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Rat | |
CA6, CA-6, Car6, Carbonate dehydratase VI, carbonic anhydrase 6, Carbonic anhydrase VI, carbonic anhydrase VI nirs variant 2, Carbonic anhydrase6, CA-VI, DOC1, GUSTIN, salivary carbonic anhydrase, Secreted carbonic anhydrase | |
Car6 | |
IgG | |
Protein A | |
Car6 |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300, pH 7.4 | |
298657 | |
Recombinant Rat CA6 protein, Thr147-Ala275 (Accession #F1LQ08) | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title