Learn More
CD5 Monoclonal Antibody (UCHT2), eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
Supplier: LIFE TECHNOLOGIES 14005995

Description
The UCHT2 monoclonal antibody reacts with human CD5, a 67 kDa protein expressed by a majority of thymocytes and mature T cells and a subset of B cells. Signaling through the CD5 molecule activates T cells and binding of CD5 to its ligand on B cells, CD72, and plays an important role in T-B interaction and proliferation. The monoclonal antibody UCHT2 recognizes primate CD5. Applications Reported: The UCHT2 antibody has been reported for use in flow cytometric analysis, immunoblotting (WB), and immunohistochemical staining. Applications Tested: The UCHT2 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
CD5 Monoclonal antibody specifically detects CD5 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen), Western BlotSpecifications
CD5 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P06127 | |
Mouse | |
10 mg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Flow Cytometry, Immunohistochemistry (Frozen), Western Blot | |
UCHT2 | |
Unconjugated | |
Cd5 | |
CD antigen CD5; CD5; CD5 antigen; CD5 antigen (p56-62); CD5 antigen p56-62; Cd5 molecule; CD5 protein; CD74 antigen, invariant polypeptide of major; fCD5; LEU1; Ly-1; Ly-12; Ly-A; Lymphocyte antigen 1; Lymphocyte antigen CD5; lymphocyte antigen T1/Leu-1; Lyt-1; Pan T cell; T1; T-cell surface glycoprotein CD5; unnamed protein product | |
Affinity chromatography | |
RUO | |
921 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.