Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CpG oligodeoxynucleotides (2216), Novus Biologicals™
Shop All Novus Biologicals Products

Click to view available options
Quantity:
0.025 mg
0.1 mg
Description
Immunogen: G&asterisk;GGGGACGATCGTCG&asterisk;G&asterisk;G&asterisk;G&asterisk;G&asterisk;G Buffer: Sterile water Clone: 2216 Dilution: Ligand Activation, 5 to 20μg/mL CpG ODN Type A (2216)
Specifications
Specifications
Concentration | 1.0 mg/mL |
Content And Storage | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Quantity | 0.1 mg |
For Use With (Application) | Human CpG ODNs are used for activation of human TLR9 at a range of 5 to 20μL. The TLR9/NF-kB/SEAPorter HEK 293 cell line is a useful positive control model system for studying hTLR9 activation. Typically use a 96-well plate format assay using this reporter cell line. |
Species | Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction