Learn More
3M™ DBI-SALA™ ExoFit NEX™ Tower Climbing Harness
3M™ DBI-SALA™ ExoFit NEX™ Tower Climbing Harness features aluminum back and front D-rings, belt with pad and side D-rings, seat sling with suspension D-rings, locking quick connect buckle leg straps, and comfort padding.
Supplier: 3M™ 7100188764
Description
- Hybrid shoulder, back and leg comfort padding
- Stand-up lightweight aluminum back D-ring
- Lightweight aluminum front D-ring
- Body belt/hip pad with lightweight aluminum side D-rings
- Removable seat sling with aluminum board and suspension D-rings
- Web-locking quick-connect chest and leg straps
- Repel™ webbing
- Built-in lanyard keepers
- Revolving vertical torso buckle
- Impact Indicator
- Integrated trauma straps
- Built-in reflective materials
- Protected labels, equipped with 3M™ Connected Safety ID (CSID)
Specifications
Duo-Lok™ Quick Connect | |
Small | |
190 kg | |
Tower Climbing Harness | |
9.7 lb. |
Repel™ Polyester | |
420 lb. | |
OSHA 1926.502, ANSI Z359.4, ANSI A10.32, OSHA 1910.66, ANSI Z359.3, ANSI Z359.1 | |
Back, Side, Front, Suspension |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.