Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ DBI-SALA™ ExoFit NEX™ Tower Climbing Harness

3M™ DBI-SALA™ ExoFit NEX™ Tower Climbing Harness features aluminum back and front D-rings, belt with pad and side D-rings, seat sling with suspension D-rings, locking quick connect buckle leg straps, and comfort padding.

$5023.83

Specifications

Buckle Style Duo-Lok™ Quick Connect
Capacity (English) 420 lb.
Certifications/Compliance OSHA 1926.502, ANSI Z359.4, ANSI A10.32, OSHA 1910.66, ANSI Z359.3, ANSI Z359.1
D-Ring Location Back, Side, Front, Suspension
Product Type Tower Climbing Harness
View More Specs

Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-809-700
View Documents
3M™
7100188764
Small
Case for $5,023.83
missing translation for 'QuantityLeft'
Add to Cart
 
19-809-725
View Documents
3M™
7100188766
Large
Case for $5,023.83
missing translation for 'QuantityLeft'
Add to Cart
 
Description

Description

  • Hybrid shoulder, back and leg comfort padding
  • Stand-up lightweight aluminum back D-ring
  • Lightweight aluminum front D-ring
  • Body belt/hip pad with lightweight aluminum side D-rings
  • Removable seat sling with aluminum board and suspension D-rings
  • Web-locking quick-connect chest and leg straps
  • Repel™ webbing
  • Built-in lanyard keepers
  • Revolving vertical torso buckle
  • Impact Indicator
  • Integrated trauma straps
  • Built-in reflective materials
  • Protected labels, equipped with 3M™ Connected Safety ID (CSID)
Specifications

Specifications

Duo-Lok™ Quick Connect
OSHA 1926.502, ANSI Z359.4, ANSI A10.32, OSHA 1910.66, ANSI Z359.3, ANSI Z359.1
Tower Climbing Harness
9.7 lb.
420 lb.
Back, Side, Front, Suspension
Repel™ Polyester
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.