Learn More
Dopamine Receptor D4 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2924180.1ML
Description
Dopamine Receptor D4 Polyclonal antibody specifically detects Dopamine Receptor D4 in Mouse samples. It is validated for Western BlotSpecifications
Dopamine Receptor D4 | |
Polyclonal | |
Western Blot 1:500-1:1000 | |
D(2C) dopamine receptor, D(4) dopamine receptor, D4DR, Dopamine D4 receptor, dopamine receptor D4, seven transmembrane helix receptor | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Dopamine Receptor D4 (P21917). MGNRSTADADGLLAGRGPAAGASAGASAGLAGQGAAALVGGVLLIGAVLAGNSLVCVSVATERALQTPTNSFIVSLAAADLLLALLVLPLFVYSEVQGGA | |
0.1 mL | |
GPCR, Neuroscience | |
1815 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.