Learn More
Dopamine Receptor D4 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$173.10 - $403.50
Specifications
Antigen | Dopamine Receptor D4 |
---|---|
Dilution | Western Blot 1:500-1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Dopamine Receptor D4 Polyclonal antibody specifically detects Dopamine Receptor D4 in Mouse samples. It is validated for Western BlotSpecifications
Dopamine Receptor D4 | |
Western Blot | |
Unconjugated | |
Rabbit | |
GPCR, Neuroscience | |
PBS with 50% glycerol, pH7.3. | |
1815 | |
IgG | |
Affinity purified |
Western Blot 1:500-1:1000 | |
Polyclonal | |
Purified | |
RUO | |
Mouse | |
D(2C) dopamine receptor, D(4) dopamine receptor, D4DR, Dopamine D4 receptor, dopamine receptor D4, seven transmembrane helix receptor | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Dopamine Receptor D4 (P21917). MGNRSTADADGLLAGRGPAAGASAGASAGLAGQGAAALVGGVLLIGAVLAGNSLVCVSVATERALQTPTNSFIVSLAAADLLLALLVLPLFVYSEVQGGA | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.