Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Wheaton™ DualFusion Vials
For storage and transport of samples. Wheaton™ DualFusion Vials maintains sample integrity and protect sensitive reagents.
Supplier: DWK Life Sciences W266104
Description
- Combines best properties of both plastic and glass into one vial
- “FUSION” material lining provides protection against leachates, delamination, and gas permeation
- Cyclic Olefin Polymer (COP) shell offers the comfort of a break resistant vial
- Manufactured in a Class 7 cleanroom so users receive a “ready-to-sterilize” vial
- Equipped with a 2D bar code which provides complete manufacturing traceability
- Can be sterilized via Gamma Radiation, ETO and E-beam
Compliance
Meet USP<788> for low particulate levels and USP<85> for low endotoxin level for injectables
Specifications
22 mm | |
5 mL | |
48 | |
Crimp Top | |
Meet USP<788> for low particulate levels and USP<85> for low endotoxin level for injecta | |
20 mm |
Clear | |
FUSION | |
-196°C to +121°C | |
40 mm | |
DualFusion Vial 6 R |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction