Learn More
DWK Life Sciences Wheaton™ DualFusion Vials
For storage and transport of samples. Wheaton™ DualFusion Vials maintains sample integrity and protect sensitive reagents.
Supplier:  DWK Life Sciences W266106
 View more versions of this product
Catalog No. 03-410-751
Description
Description
  • Combines best properties of both plastic and glass into one vial
  • “FUSION” material lining provides protection against leachates, delamination, and gas permeation
  • Cyclic Olefin Polymer (COP) shell offers the comfort of a break resistant vial
  • Manufactured in a Class 7 cleanroom so users receive a “ready-to-sterilize” vial
  • Equipped with a 2D bar code which provides complete manufacturing traceability
  • Can be sterilized via Gamma Radiation, ETO and E-beam
Compliance
Meet USP<788> for low particulate levels and USP<85> for low endotoxin level for injectables
Specifications
Specifications
22 mm
5 mL
96
Crimp Top
Meet USP<788> for low particulate levels and USP<85> for low endotoxin level for injecta
20 mm
Clear
FUSION
-196°C to +121°C
40 mm
DualFusion Vial 6 R
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.