Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ EcoPoint™ Variable Volume Micropipettes
SDPSCI_ED

EcoPoint™ Variable Volume Micropipettes offer accurate ergonomic pipetting at a great value.

Supplier:  United Scientific™ EVP200

 View more versions of this product

Catalog No. S03084


missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart

Description

Description

Our EcoPoint™ variable volume pipetters are designed to dispense liquids with a high degree of accuracy, and are very easy to work with. The lightweight plunger is designed to facilitate use for extended periods of time.

Each pipette box includes a calibrated pipetter, sample pipette tips, a calibration tool, a quality control calibration certificate, and a user's manual.

  • Available in these sizes (μL): 0.5-10, 2-20, 5-50, 10-100, 20-200, 100-1000, 1000-5000, 2000-10000
  • Designed for use with universal pipette tips (depending on the volume range)
  • Fully autoclavable (121°C, 0.15Mpa)
  • Manufactured, calibrated, and tested per ISO standards
  • CE certified
Specifications

Specifications

±0.8%
Autoclavable
Reusable
Ergonomic
1
Non-sterile
20 to 200 μL
20 to 200 μL
White
Pipette Tip 2 to 200 μL
Plastic
No
Adjustable Single Channel Pipette
1 μL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.