Learn More
Fisherbrand™ Executive Windrowe Mesh Back Chair with Grade A Seat, Head Rest, and 3D Adjustable Arms
SDP
Fisherbrand™ Executive Windrowe Mesh Back Chairs with Grade A Vinyl Seat are ideal for office environments or anywhere where seats need to be cleaned. Grade A vinyl provides additional antibacterial protections. Head rest and 3D adjustable arms offer adjustments to ensure proper arm placement.
 View more versions of this product
Catalog No. 15-001-969
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Description
Description
  • Adjustable from 19 to 23 inches from the top of the seat to the floor
  • Features adjustable lumbar support
  • 3D adjustable arm rests adjust up and down, twist toward and away from the user, and slide forward the back
  • Comfort molded seat
  • Head rest
  • Composite base
  • Antifungal/antibacterial Grade A Vinyl
  • Additional antibacterial protections
  • High end look and feel
  • Variety of color options to suit any work space
Specifications
Specifications
3D adjustable arm rests move up and down-in and out-forward and backward-and twist towards and away from user.
Carbon
No
Supernova Grade A Vinyl
Desk Height
Yes
No
Nylon Reinforced Fiberglass Base
Black dual-wheel, casters (suitable for carpeted floors)
Round tube chrome foot ring with internal lock
Office/General Seating
64 lb.
Yes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.