Learn More
Jackson Safety Face Shield Cap Adapters
Jackson Safety 282 B Cord Mounted Cap Adapters for Non-Slotted Caps features a polyethylene spark deflector. These cord-mounted, cap adapters, designed specifically for non-slotted caps are available with face shields (sold separately), that are secured with internal plastic lugs.
$483.00 - $1388.00
Specifications
Color Silver
Size Universal
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Equipment) Mounting Type Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Mounting Type Price Quantity & Availability  
19-060-785
View Documents
Jackson Safety
14943
Hard Hats Metal Clips
Case for $483.00
Add to Cart
19-060-791
View Documents
Jackson Safety
14952
Slotted Hard Hat,Universal Faceshield Windows Slotted
Case for $1,388.00
Add to Cart
Description
Description
  • Model 282-B Cap Adaptor with Crown
  • Vertical lock-in eliminates face shield creep
  • Oversized crown for added safety
  • Specially designed metal clips press over the hard hat rim for maximum stability and firm fit
  • Features five easy-to-swivel lock mounting pins for universal windows
  • An extremely popular cap adaptor
Specifications
Specifications
Silver
Universal
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.