Learn More
Fehling’s Solution A Copper Sulfate Solution, Reagents
Fehling’s Solution A Copper Sulfate Solution, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc C720200020F
Specifications
Fehling’s Solution A Copper Sulfate Solution | |
7732-18-5, 7758-99-8 | |
Reagent | |
1.04 | |
Non-sterile |
Blue | |
100°C | |
Cubitainer | |
20 L | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.