Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fehling’s Solution A Copper Sulfate Solution, Reagents

Fehling’s Solution A Copper Sulfate Solution, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

$33.45 - $319.50

Specifications

Chemical Name or Material Fehling’s Solution A Copper Sulfate Solution
Color Blue
CAS 7732-18-5, 7758-99-8
Boiling Point 100°C
Grade Reagent
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
01-336-496
View Documents
Reagents Holdings Llc
C72020001A
1 L
Each for $50.70
missing translation for 'QuantityLeft'
Add to Cart
 
01-336-495
View Documents
Reagents Holdings Llc
C720200020F
20 L
Each for $319.50
missing translation for 'QuantityLeft'
Add to Cart
 
01-336-494
View Documents
Reagents Holdings Llc
C72020004A
4 L
Each for $106.50
missing translation for 'QuantityLeft'
Add to Cart
 
01-336-493
View Documents
Reagents Holdings Llc
C7202000500A
500 mL
Each for $33.45
missing translation for 'QuantityLeft'
Add to Cart
 
Description

Description

Specifications

Specifications

Fehling’s Solution A Copper Sulfate Solution
7732-18-5, 7758-99-8
Reagent
Non-sterile
Blue
100°C
1.04
Liquid
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.