Learn More
Hamilton™ Gastight™ Syringe, 100 μL, Sold by MilliporeSigma™ Supelco™
Gas-tight design minimizes gas exchange during GC analysis. Syringes provide accurate volume delivery with a plunger that is mostly unsurpassed.
Supplier:  MilliporeSigma Supelco 26205
 View more versions of this product
Catalog No. 11-101-1169
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Description
Description
Specifications
Specifications
Autoclavable
Gastight™ Syringe
239°F
50°F
10°C
100 μL
PTFE Luer Lock (TLL) with Slots
Borosilicate Glass (Barrel), PTFE (Plunger)
1000 psig (68.9 bar)
115°C
1710 TLL
1 Ea.
Hamilton
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
GASTIGHT is a registered trademark of Hamilton Co.