Learn More
HMGXB4 Rabbit anti-Human, Proteintech
SDP
Rabbit PolyClonal Antibody
Supplier:  Proteintech Group Inc 248211AP150UL
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-4561
missing translation for 'QuantityLeft'
Add to Cart
Description
Description
High mobility group (HMG) proteins are nonhistone chromosomal proteins.
Specifications
Specifications
HMGXB4
Polyclonal
Unconjugated
HMGXB4
HMG box containing protein 4, HMG box domain containing 4, HMG2L1, HMGBCG, HMGXB4, Protein HMGBCG, THC211630
Rabbit
Antigen Affinity Chromatography
RUO
10042
-20°C
Liquid
Immunoprecipitation, Western Blot
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9UGU5
HMGXB4
HMGXB4 Fusion Protein Ag20424
150 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions