Learn More
HMGXB4 Rabbit anti-Human, Proteintech

Rabbit PolyClonal Antibody
$516.35
Specifications
Antigen | HMGXB4 |
---|---|
Concentration | 0.37 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-4561
![]() |
Proteintech Group Inc
248211AP150UL |
150 μL |
Each of 1 for $516.35
|
|
|||||
Description
High mobility group (HMG) proteins are nonhistone chromosomal proteins.Specifications
HMGXB4 | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Antigen Affinity Chromatography |
0.37 mg/mL | |
Polyclonal | |
Rabbit | |
Human | |
IgG |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.