Learn More
Diazyme Laboratories Homocysteine Enzymatic Assay
Liquid stable system that provides a fast, cost effective alternative to immunochemical methods
Supplier:  Diazyme Laboratories DZ568AK03
 View more versions of this product
Catalog No. 23-700-644
This item is not returnable. View return policy
Description
Description
  • No significant interference from lipemic and hemolytic substances
  • Eliminates carry over interference with iron or lipase reagents
  • Assay not interfered by endogenous cystathionine
  • For in vitro quantitative determination of total L-homocysteine in serum or plasma
  • Assists in diagnosis and treatment of patients suspected of having hyperhomocysteinemia and homocystinuria
Specifications
Specifications
510(k) cleared, Health Canada registered
Yes
R1: 1 x 25.5 mL, R2: 1 x 5.8 mL, R3: 1 x 4.8 mL
Roche™ Hitachi, Siemens Dimension™ and Beckman Synchron CX, LX and DXC analyzers
3 to 50 μmol/L
Homocysteine Enzymatic Assay
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.