Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diazyme Laboratories Homocysteine Enzymatic Assay
Liquid stable system that provides a fast, cost effective alternative to immunochemical methods
$2460.75
Specifications
Certifications/Compliance | 510(k) cleared, Health Canada registered |
---|---|
CE Marker | Yes |
Type | Homocysteine Enzymatic Assay |
Linearity | 3 to 50 μmol/L |
Form | Liquid |
Catalog Number | Mfr. No. | For Use With (Equipment) | Form | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Form | Includes | Price | Quantity & Availability | |||
23-700-645
![]() |
Diazyme Laboratories
DZ568BBY2 |
Beckman AU (400/600/640/680) analyzers | Liquid |
Each of 1 for $2,460.75
|
|
||||
23-700-644
|
Diazyme Laboratories
DZ568AK03 |
Roche™ Hitachi, Siemens Dimension™ and Beckman Synchron CX, LX and DXC analyzers | N/A | N/A | N/A | ||||
Description
- No significant interference from lipemic and hemolytic substances
- Eliminates carry over interference with iron or lipase reagents
- Assay not interfered by endogenous cystathionine
- For in vitro quantitative determination of total L-homocysteine in serum or plasma
- Assists in diagnosis and treatment of patients suspected of having hyperhomocysteinemia and homocystinuria
Specifications
510(k) cleared, Health Canada registered | |
Homocysteine Enzymatic Assay | |
Liquid |
Yes | |
3 to 50 μmol/L |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title