Learn More
Novus Biologicals™ Human Fallopian Tube Whole Tissue Lysate (Adult Whole Normal)
Shop All Novus Biologicals Products

Specifications
For Use With (Application)
Western Blot
Tissue
Reproductive
Quantity
1 mg
Storage Buffer
HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Test Specificity
Total Protein - Human Adult Normal Tissue: Fallopian Tube
Cellular Fraction
Whole
Tissue Condition
Normal
Cross Reactivity
Human
Host Species
Human
Concentration
5 mg/mL
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.