Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Heart Interventricular Septum Whole Tissue Lysate (Adult Whole Normal)
SDP

Catalog No. NB82059223 Shop All Novus Biologicals Products
Click to view available options
:
The Human Heart Interventricular Septum Whole Tissue Lysate (Adult Whole Normal) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.

Specifications

For Use With (Application)

Western Blot

Tissue

Heart

Quantity

1 mg

Storage Buffer

HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors

Test Specificity

Total Protein - Human Adult Normal Tissue: Heart: Interventricular Septum

Cellular Fraction

Whole

Tissue Condition

Normal

Cross Reactivity

Human

Host Species

Human

Concentration

5 mg/mL

Product Type

Human Heart Interventricular Septum Whole Tissue Lysate (Adult Normal)

Content And Storage

-80°C. Avoid freeze-thaw cycles.

Life Stage

Adult

Protein State

Native

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.