Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Human Heart Pericardium Whole Tissue Lysate (Adult Whole Normal)
Shop All Novus Biologicals Products

Click to view available options
:
1mg
Description
The Human Heart Pericardium Whole Tissue Lysate (Adult Whole Normal) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.
Specifications
Specifications
For Use With (Application) | Western Blot |
Tissue | Heart |
Quantity | 1 mg |
Storage Buffer | HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors |
Test Specificity | Total Protein - Human Adult Normal Tissue: Heart: Pericardium |
Cellular Fraction | Whole |
Tissue Condition | Normal |
Cross Reactivity | Human |
Host Species | Human |
Concentration | 1 mg/mL |
Show More |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction