Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydrogen Potassium ATPase Beta Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP19157320UL
Description
Hydrogen Potassium ATPase Beta Polyclonal specifically detects Hydrogen Potassium ATPase Beta in Rat samples. It is validated for Western Blot.Specifications
Hydrogen Potassium ATPase Beta | |
Polyclonal | |
Western Blot 0.2-1 ug/ml | |
NP_036642 | |
ATP4B | |
Synthetic peptide directed towards the C terminal of human Atp4b. Peptide sequence QPHYSNPLVAAKFLNVPKNTQVLIVCKIMADHVTFDNPHDPYEGKVEFKL. | |
20 μL | |
Primary | |
Rat | |
IgG |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
ATP6B, ATPase, H+/K+ exchanging, beta polypeptide, ATPase, H+/K+ transporting, beta polypeptide, Gastric H(+)/K(+) ATPase subunit beta, gastric H+/K+ ATPase beta subunit, gastric hydrogen-potassium ATPase, beta, potassium-transporting ATPase beta chain, potassium-transporting ATPase subunit beta, Proton pump beta chain | |
Rabbit | |
Affinity Purified | |
RUO | |
496 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?