Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydrogen Potassium ATPase Beta Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$165.00 - $487.50
Specifications
Antigen | Hydrogen Potassium ATPase Beta |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP191573
![]() |
Novus Biologicals
NBP191573 |
100 μL |
Each for $487.50
|
|
|||||
NBP19157320
![]() |
Novus Biologicals
NBP19157320UL |
20 μL |
Each for $165.00
|
|
|||||
Description
Hydrogen Potassium ATPase Beta Polyclonal specifically detects Hydrogen Potassium ATPase Beta in Rat samples. It is validated for Western Blot.Specifications
Hydrogen Potassium ATPase Beta | |
Polyclonal | |
Rabbit | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
ATP6B, ATPase, H+/K+ exchanging, beta polypeptide, ATPase, H+/K+ transporting, beta polypeptide, Gastric H(+)/K(+) ATPase subunit beta, gastric H+/K+ ATPase beta subunit, gastric hydrogen-potassium ATPase, beta, potassium-transporting ATPase beta chain, potassium-transporting ATPase subunit beta, Proton pump beta chain | |
ATP4B | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
RUO | |
NP_036642 | |
496 | |
Synthetic peptide directed towards the C terminal of human Atp4b. Peptide sequence QPHYSNPLVAAKFLNVPKNTQVLIVCKIMADHVTFDNPHDPYEGKVEFKL. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title