Learn More
ISOC2, Polyclonal, Bethyl Laboratories

Description
The recommended shelf life for this product is 1 year from date of receipt.
ISOC2 gene ontology annotations related to this gene include protein destabilization.Specifications
ISOC2 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q96AB3 | |
ISOC2 | |
Between 75 and 125 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ISOC2 | |
Isochorismatase domain-containing protein 2, mitochondrial, ISOC2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79763 | |
4° C | |
Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.