Learn More
ISOC2, Polyclonal, Bethyl Laboratories

Rabbit Polyclonal Antibody
$154.46
Specifications
Classification | Polyclonal |
---|---|
Primary or Secondary | Primary |
Antigen | ISOC2 |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1151
![]() |
Bethyl Laboratories, Inc
A305754AT |
20 μL |
Each of 1 for $154.46
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
ISOC2 gene ontology annotations related to this gene include protein destabilization.Specifications
Polyclonal | |
Primary | |
ISOC2 | |
Unconjugated |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.