Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ ITD 1
Selective inhibitor of TGF-β signaling; induces cardiomyocyte differentiation in ESCs
$342.50 - $1396.00
Specifications
Purity | >98% |
---|---|
Content And Storage | -20°C |
Product Type | ITD 1 |
Molecular Weight (g/mol) | 415.52 |
Inhibitors | ITD 1 |
Description
Selective inhibitor of TGF-β signaling (IC50 = 0.85 μM); displays little or no inhibition of activin, Wnt or BMP signaling pathways. Induces proteasomal degradation of the TGF-β type II receptor. Inhibits TGF-β-induced mesoderm formation from mouse embryonic stem cells (ESCs) during early differentiation; selectively promotes the differentiation of ESCs to cardiomyocytes in vitro between days 3-5. Does not induce differentiation of vascular smooth muscle cells or endothelial cells.Specifications
>98% | |
ITD 1 | |
ITD 1 |
-20°C | |
415.52 |
For Research Use Only
Spot an opportunity for improvement?
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.