Learn More
Novus Biologicals™ L929 CoCl2 treated/Untreated Cell Lysate
Shop All Novus Biologicals Products

Specifications
Preparation Method
L929 whole cell lysate is provided as a Western blot positive control. The lysates are provided as a set of treated and untreated samples. Use 10 μL / lane for a standard
Quantity
2 Vials
Host Species
Mouse
Content And Storage
Store at -20°C to -80°C. Avoid freeze/thaw cycles.
Target
L929
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.