Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LabAid Universal Viral Collection Kit
Designed to collect and transport viruses, chlamydiae, mycoplasmas and urea plasmas at ambient temperature.
Supplier: Biomed Resources Inc BVTK003
This item is not returnable.
View return policy
Description
- Combines a flexible minitip collection swab and vial for collection
- Can be stored and transported at 0°C to 25°C - all in one formulation
- Maintains viability of organisms - antibiotics inhibit bacteria and fungal flora
- Maintains microbial viability for safe transport
- Cost efficient large 3 mL media fill volume allows for multiple tests on the same specimen
- Multi-purpose usage with design variations
- Consists of high density materials which meet stringent QC standards
Specifications
Flexible Minitip Flocked Swab | |
1,600 nasal swabs and 1,600 5 mL tubes with 3 mL inactivated specimen preservation solution | |
Flexible Minitip | |
5 mL |
Orange | |
0°C to 25°C | |
0-25°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction