Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Liver Matched Tumor and Normal Tissue Lysate
Shop All Novus Biologicals Products

Click to view available options
:
Diagnosis: Liver tumor
Description
The Liver Matched Tumor & Normal Tissue Lysate is derived from human.
Specifications
Specifications
Tissue | Liver |
Quantity | 2 Vials |
Host Species | Human |
Content And Storage | Store at −80°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction