Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Maxi Potassium channel alpha Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

Supplier:  Novus Biologicals NBP1467010.025ML

 View more versions of this product

Catalog No. NB018638


Explore all special offers
Add to Cart

Description

Description

Maxi Potassium channel alpha Polyclonal specifically detects Maxi Potassium channel alpha in Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Maxi Potassium channel alpha
Polyclonal
Unconjugated
Whole antisera with No Preservative
bA205K10.1, BK channel, BKCA alpha, BKCA alpha subunit, BKTM, calcium-activated potassium channel subunit alpha-1, DKFZp686K1437, hSlo, k(VCA)alpha, KCNMA, Maxi K channel, MaxiK, MGC71881, potassium large conductance calcium-activated channel, subfamily M, alphamember 1, SAKCA, Slo homolog, Slo1, SLO-ALPHA, Slowpoke homolog, stretch-activated Kca channel, subfamily M subunit alpha-1
Rabbit
Unpurified
RUO
Primary
Rat, mouse. Other species not yet tested.
Mouse, Rat
Antisera
Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot
LYOPH
Western Blot 1:300-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:300-1:2000
Q62976
KCNMA1
A synthetic peptide from 4th cytoplasmic domain of rat Potassium channel subfamily M subunit alpha 1 (KCNMA1, KCa1.1, SLO) conjugated to an immunogenic carrier protein was used as the antigen.
0.025 mL
Potassium Channels
3778
Reconstitute with 0.1 ml sterilized water. Centrifuge to remove any insoluble material.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only