Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Maxi Potassium channel alpha Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

$181.90 - $487.50

Specifications

Antigen Maxi Potassium channel alpha
Concentration LYOPH
Classification Polyclonal
Conjugate Unconjugated
Form Antisera
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018638
SDP
View Documents
Novus Biologicals
NBP1467010.025ML
0.025 mL
Each for $181.90
Add to Cart
 
NBP146701
SDP
View Documents
Novus Biologicals
NBP146701
0.1 mL
Each for $487.50
Add to Cart
 
Description

Description

Maxi Potassium channel alpha Polyclonal specifically detects Maxi Potassium channel alpha in Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Maxi Potassium channel alpha
Polyclonal
Antisera
RUO
Q62976
3778
IgG
Unpurified
Rat, mouse. Other species not yet tested.
LYOPH
Unconjugated
Rabbit
Potassium Channels
bA205K10.1, BK channel, BKCA alpha, BKCA alpha subunit, BKTM, calcium-activated potassium channel subunit alpha-1, DKFZp686K1437, hSlo, k(VCA)alpha, KCNMA, Maxi K channel, MaxiK, MGC71881, potassium large conductance calcium-activated channel, subfamily M, alphamember 1, SAKCA, Slo homolog, Slo1, SLO-ALPHA, Slowpoke homolog, stretch-activated Kca channel, subfamily M subunit alpha-1
KCNMA1
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.