Learn More
Maxi Potassium channel alpha Antibody, Novus Biologicals™
SDP
Rabbit Polyclonal Antibody has been used in 1 publication
Supplier:  Novus Biologicals NBP146701
 View more versions of this product
Catalog No. NBP146701
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Description
Description
Maxi Potassium channel alpha Polyclonal antibody specifically detects Maxi Potassium channel alpha in Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications
Specifications
Maxi Potassium channel alpha
Polyclonal
Unconjugated
Lyophilized from whole antisera with No Preservative
bA205K10.1, BK channel, BKCA alpha, BKCA alpha subunit, BKTM, calcium-activated potassium channel subunit alpha-1, DKFZp686K1437, hSlo, k(VCA)alpha, KCNMA, Maxi K channel, MaxiK, MGC71881, potassium large conductance calcium-activated channel, subfamily M, alphamember 1, SAKCA, Slo homolog, Slo1, SLO-ALPHA, Slowpoke homolog, stretch-activated Kca channel, subfamily M subunit alpha-1
Rabbit
Unpurified
RUO
Primary
Rat, mouse. Other species not yet tested.
Mouse, Rat, Monkey
Antisera
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
LYOPH
Western Blot 1:2000, Flow Cytometry, Immunohistochemistry 1:2000, Immunohistochemistry-Paraffin 1:2000
Q62976
KCNMA1
A synthetic peptide from rat Maxi Potassium channel alpha conjugated to blue carrier protein was used as the antigen. The peptide is homologous in mouse.
0.1 mL
Potassium Channels
3778
Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size is provided in reconstituted format.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only