Learn More
MCR Safety Indirect Vent Goggles
The body design of the Verdict™ provides a wide, unobstructed field of vision and can be worn with most half-mask respirators. Transparent protective PVC body permits a comfortable fit over prescription eyewear, and the optional soft foam lining cushions the face and improves the seal.
Supplier: MCR Safety 2400
Description
Offered in both scratch-resistant polycarbonate lens and anti-fog lens, the Verdict is also available with either a wide elastic headband or a neoprene headband.- Impact Resistant
- Fits over Prescription Glasses
- Optional Foam Lining
- Indirect Ventilation
- ANSI Z87+ and CSA Z94.3
Compliance
Meets ANSI Z87.1-2015, ANSI D3, ANSI D4 and CSA Z94.3 standards.
Specifications
Scratch-resistant | |
Clear | |
Smoke | |
Venting is indirect | |
PVC |
ANSI Z87.1 | |
Safety Goggles | |
Wide elastic headband | |
Polycarbonate |