Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PrimeStore™ MTM Molecular Transport Media
SDP

PrimeStore™ MTM is a FDA Cleared Microbial nucleic acid storage and stabilization device. It inactivates microbes (to include SARS-CoV-2), stabilizes RNA/DNA and is on the US FDA list of primary sample collection devices for SARA-CoV-2 molecular testing.

Supplier:  PrimeStore™ MTM LH105CC

 View more versions of this product

Catalog No. 23-100-403


Explore all special offers
Add to Cart
This item is not returnable. View return policy

Description

Description

  • PrimeStore™ MTM is the only FDA Cleared (cleared in 2018) Molecular Transport Medium for respiratory samples suspected of harboring influenza or TB.
  • Considered by US FDA as a primary sample collection device for SARS-CoV-2.
  • PrimeStore™ collected samples enable safe handling for both point of care and in the laboratory--since the entire sample in inactivated/killed including SARS-CoV-2 and more resilient microbes like gram positives, etc.
  • Once a swab or biofluid is added to PrimeStore™ the user has a “snapshot” in time for that sample.
  • The DNA, and more importantly the labile RNA is preserved/stabilized.
  • Groups all over the United States, Europe, and Africa/Asia use PrimeStore™ to move and transport samples across borders using NORMAL FedEX and UPS shipping.
    • Since the pathogens collected are eliminated, the samples can be shipped using routine boxing/shipping methods.
    • Samples in PrimeStore™ can be shipped with NO COLD CHAIN, no wet or dry ice.
  • Samples do not have to be pre-processed in BLS-III or even BLS-II safety levels.
  • Safe or normal technician testing.
  • The collected sample in PrimeStore™ can be used/re-used several times for additional testing.
  • Many Labs use the first sampling for detection by qPCR and the second for NGS or gene sequencing.
  • PrimeStore™ MTM was designed to be compatible with all Qiagen-based extraction systems (bench top and high throughput liquid handling systems). It works with the Thermo Scientific MagMax, Roche MAGNAPURE or almost all DNA/RNA liquid handling extraction systems.
  • The medium if in cryotubes can be frozen at ultra-low for biobanking or simply stored in the refrigerator (since the medium is safely preserving the collected sample).
Specifications

Specifications

Molecular Testing-Clinical
50 tubes per box
Ambient
1.5 mL
5 mL
Wide mouth preform tube
5 mL wide mouth preform tubes with 1.5 mL of PrimeStore™ MTM reagent
Ambient
FDA Cleared
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 2 years

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.