Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PrimeStore™ MTM Molecular Transport Media

PrimeStore™ MTM is a FDA Cleared Microbial nucleic acid storage and stabilization device. It inactivates microbes (to include SARS-CoV-2), stabilizes RNA/DNA and is on the US FDA list of primary sample collection devices for SARA-CoV-2 molecular testing.
$186.67 - $558.50
Specifications
Certifications/Compliance | FDA Cleared |
---|---|
Color | Clear |
For Use With (Application) | Molecular Testing-Clinical |
Quantity | 50 Tubes/Pk. |
Storage Requirements | Ambient |
Catalog Number | Mfr. No. | Fill Volume | Tube Size | Packaging | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Fill Volume | Tube Size | Packaging | Price | Quantity & Availability | |||
23-100-400
![]() |
PrimeStore™ MTM
LH102 |
1 mL | Cryotube |
Pack of 50 for $186.67
|
|
||||
23-100-401
![]() |
PrimeStore™ MTM
LH103 |
1.5 mL | Cryotube |
Pack of 50 for $257.50
|
|
||||
23-100-402
![]() |
PrimeStore™ MTM
LH105 |
1.5 mL | Cryotube |
Pack of 50 for $257.50
|
|
||||
23-100-403
![]() |
PrimeStore™ MTM
LH105CC |
1.5 mL | 5 mL | Wide mouth preform tube |
Pack of 50 for $558.50
|
|
|||
Description
- PrimeStore™ MTM is the only FDA Cleared (cleared in 2018) Molecular Transport Medium for respiratory samples suspected of harboring influenza or TB.
- Considered by US FDA as a primary sample collection device for SARS-CoV-2.
- PrimeStore™ collected samples enable safe handling for both point of care and in the laboratory--since the entire sample in inactivated/killed including SARS-CoV-2 and more resilient microbes like gram positives, etc.
- Once a swab or biofluid is added to PrimeStore™ the user has a “snapshot” in time for that sample.
- The DNA, and more importantly the labile RNA is preserved/stabilized.
- Groups all over the United States, Europe, and Africa/Asia use PrimeStore™ to move and transport samples across borders using NORMAL FedEX and UPS shipping.
- Since the pathogens collected are eliminated, the samples can be shipped using routine boxing/shipping methods.
- Samples in PrimeStore™ can be shipped with NO COLD CHAIN, no wet or dry ice.
- Samples do not have to be pre-processed in BLS-III or even BLS-II safety levels.
- Safe or normal technician testing.
- The collected sample in PrimeStore™ can be used/re-used several times for additional testing.
- Many Labs use the first sampling for detection by qPCR and the second for NGS or gene sequencing.
- PrimeStore™ MTM was designed to be compatible with all Qiagen-based extraction systems (bench top and high throughput liquid handling systems). It works with the Thermo Scientific MagMax, Roche MAGNAPURE or almost all DNA/RNA liquid handling extraction systems.
- The medium if in cryotubes can be frozen at ultra-low for biobanking or simply stored in the refrigerator (since the medium is safely preserving the collected sample).
Specifications
FDA Cleared | |
Molecular Testing-Clinical | |
Ambient |
Clear | |
50 Tubes/Pk. |
Safety and Handling
ShelfLife : 2 Years
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title