Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Adenosine Deaminase/ADA Recombinant Protein
SDP

Catalog No. NBP17236250 Shop All Novus Biologicals Products
Click to view available options
:
50μg

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-363 of Human Adenosine Deaminase/ADA The Recombinant Human Adenosine Deaminase/ADA Protein is derived from E. coli. The Recombinant Human Adenosine Deaminase/ADA Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

Accession Number NP_000013
For Use With (Application) Blocking/Neutralizing, SDS-PAGE
Formulation Liquid. 20mM Tris-HCl buffer (pH8.0) containing 20% glycerol, 1mM DTT
Gene ID (Entrez) 100
Molecular Weight (g/mol) 42.9kDa
Name Adenosine Deaminase/ADA Protein
Quantity 50 μg
Source E. Coli
Immunogen 1-363aaa, Human, His tagged
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >85%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.