Learn More
Novus Biologicals™ Ovary Granulosa Cell Tissue Slides (Tumor)
Shop All Novus Biologicals Products

Specifications
Cross Reactivity
Human
Type
Ovary Granulosa Cell Tissue Slides (Tumor)
Regulatory Status
This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Species
Human
Tissue Condition
Tumor
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.