Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ p53 Antibody Pair [HRP]

Mouse Monoclonal Antibody Pair
Supplier: Novus Biologicals™ NBP2796585PLATES
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Cynomolgus Monkey p53.Specifications
156.25 pg/mL | |
Sandwich ELISA | |
Conjugated | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Cynomolgus Monkey p53. | |
Sandwich ELISA | |
7157 | |
Mouse | |
5 Plates | |
Apoptosis, Cancer, Cell Cycle and Replication, Cellular Markers, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission, p53 Pathway, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors | |
p53 |
156.25-10000 pg/mL | |
HRP | |
Antibody Pairs | |
Cynomolgus Monkey | |
Antigen NY-CO-13, FLJ92943, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, transformation-related protein 53, tumor protein p53, Tumor suppressor p53 | |
TP53 | |
RUO | |
Storage is content dependent. | |
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 1 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 10000 pg/mL is recommended. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction