Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ p53 Antibody Pair [HRP]
SDP

Mouse Monoclonal Antibody Pair

$1195.00

Specifications

Assay Range 156.25 pg/mL
Assay Sensitivity 156.25-10000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB010355
SDP
View Documents
Novus Biologicals™
NBP27965815PLATES
15 Plates
Each for $1,195.00
missing translation for 'QuantityLeft'
Add to Cart
 
NB010356
SDP
View Documents
Novus Biologicals™
NBP2796585PLATES
5 Plates N/A N/A N/A
Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Cynomolgus Monkey p53.
Specifications

Specifications

156.25 pg/mL
Sandwich ELISA
Sandwich ELISA Kit
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Cynomolgus Monkey p53.
Sandwich ELISA
7157
Mouse
Apoptosis, Cancer, Cell Cycle and Replication, Cellular Markers, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission, p53 Pathway, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors
p53
156.25-10000 pg/mL
HRP
Antibody Pairs
Cynomolgus Monkey
Antigen NY-CO-13, FLJ92943, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, transformation-related protein 53, tumor protein p53, Tumor suppressor p53
TP53
RUO
Storage is content dependent.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.