Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Parotid Tissue Slides (Normal)
SDP

Catalog No. NBP230192 Shop All Novus Biologicals Products
Click to view available options
:

Specifications

Type

Parotid Tissue Slides (Normal)

For Use With (Application)

Immunohistochemistry-Paraffin

Species

Human

Cross Reactivity

Human

Tissue Condition

Normal

Regulatory Status

This product is for research use only and is not approved for use in humans or in clinical diagnosis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.