Learn More
Novus Biologicals™ Potassium Channel Kv3.1 Antibody, Novus Biologicals™
SDP
Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals™ NBP33082425UG
 View more versions of this product
Catalog No. NB463042
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Description
Description
Potassium Channel Kv3.1 Polyclonal antibody specifically detects Potassium Channel Kv3.1 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
Specifications
Potassium Channel Kv3.1
Polyclonal
Western Blot : 5-10 μg/mL
FLJ41162, FLJ42249, FLJ43491, KV3.1, KV4, MGC129855, NGK2, potassium voltage-gated channel subfamily C member 1, potassium voltage-gated channel, Shaw-related subfamily, member 1, voltage-gated potassium channel protein KV3.1, Voltage-gated potassium channel subunit Kv3.1, Voltage-gated potassium channel subunit Kv4
Rabbits were immunized with a synthetic peptide derived from the rat Potassium Channel Kv3.1 conjugated to KLH
25 μg
Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Potassium Channels
3746
Store at -20°C. Avoid freeze/thaw cycles.
IgG
Western Blot
Unconjugated
Phosphate buffered saline
Rabbit
Ammonium sulfate precipitation
RUO
Primary
Human, Mouse, Rat
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.