Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Potassium Channel Kv3.1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$177.45 - $471.45
Specifications
Antigen | Potassium Channel Kv3.1 |
---|---|
Dilution | Western Blot : 5-10 μg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Potassium Channel Kv3.1 Polyclonal antibody specifically detects Potassium Channel Kv3.1 in Human,Mouse,Rat samples. It is validated for Western BlotSpecifications
Potassium Channel Kv3.1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Potassium Channels | |
Phosphate buffered saline | |
3746 | |
IgG | |
Ammonium sulfate precipitation |
Western Blot : 5-10 μg/mL | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
FLJ41162, FLJ42249, FLJ43491, KV3.1, KV4, MGC129855, NGK2, potassium voltage-gated channel subfamily C member 1, potassium voltage-gated channel, Shaw-related subfamily, member 1, voltage-gated potassium channel protein KV3.1, Voltage-gated potassium channel subunit Kv3.1, Voltage-gated potassium channel subunit Kv4 | |
Rabbits were immunized with a synthetic peptide derived from the rat Potassium Channel Kv3.1 conjugated to KLH | |
Primary | |
Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title