Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G-Biosciences Probe-Based qPCR 2X Master Mix

Probe-based PCR 2X master mix is used for gene expression analysis, genotyping and mutation detection, pathogen detection, viral detection, absolute quantification. It has enhanced efficiency, specificity, sensitivity and compatible with nearly all real time PCR instruments.
$80.00 - $350.00
Specifications
Detection Method | Primer-probe |
---|---|
Form | Liquid |
GC-Rich PCR Performance | High |
PCR Method | 1-step RT-qPCR, 2-step RT-qPCR, Multiplex qPCR, qPCR, RT-qPCR |
Reaction Speed | Fast or Standard |
Description
- G-biosciences probe-based qPCR 2X master mix contains Taq DNA polymerase, low ROX reference dye, dNTPs and optimized buffer with stabilizers.
- This master mix is ideal for high-throughput real-time PCR screening and validation which requires minimal handling and offer robust and consistent results.
Specifications
Primer-probe | |
High | |
Fast or Standard | |
Gene Expression, Genotyping, Microbial Detection, Pathogen Detection, Virus Detection |
Liquid | |
1-step RT-qPCR, 2-step RT-qPCR, Multiplex qPCR, qPCR, RT-qPCR | |
qPCR Master Mix |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title