Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Proteome Purify 2 Human Serum Protein Immunodepletion Resin
Shop All R&D Systems Products
Click to view available options
Quantity:
20 Tests
40 Tests
Description
Includes
Immunodepletion Resin, Spin-X Filter Units
Allows user to increase protein load or concentration
Removal allows visualization of proteins that co-migrate or co-purify with the high-abundance proteins
Kit removes proteins representing approximately 60% of the protein contained in serum and plasma
Specifications
Specifications
Quantity | 20 Tests |
Product Type | Immunodepletion Resin |
Content And Storage | Store the unopened product at 2 - 8° C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction